Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
D-Limonene, Food Grade, 95-97.5%, Spectrum™ Chemical
Shop All Spectrum Chemical Manufacturing Corporation Products

Click to view available options
Quantity:
500 g
2.5 kg
15 kg
Packaging:
Amber Glass Bottle
Description
Spectrum™ Chemical D-Limonene, Food Grade , also known as 4-Isopropenyl-1-methylcyclohexene, is used as a flavorant and as a fragrance in perfumery, aftershave lotions, bath products and other such products. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.
Specifications
Specifications
| CAS Max % | 100% |
| Boiling Point | 175°C to 176°C |
| Melting Range | -95.5°C to -74°C |
| Packaging | Amber Glass Bottle |
| Refractive Index | 1.471-1.474 |
| Quantity | 15 kg |
| UN Number | UN2052 |
| Optical Rotation | +96.0° to +104° |
| Percent Purity | 95 to 97.5% |
| Grade | Food Grade |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction