Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-ribulose, 99.7%, For HPLC analysis, MP Biomedicals™

Catalog No. ICN15202825
Click to view available options
Quantity:
10 mg
25 mg
100 mg
500 mg

Chemical Identifiers

CAS 488-84-6
Molecular Formula C5H10O5
Molecular Weight (g/mol) 150.13
InChI Key ZAQJHHRNXZUBTE-NQXXGFSBSA-N
Synonym d-ribulose, ribulose, d-erythro-pent-2-ulose, d-erythropentulose, d-erythro-pentulose, unii-z7u4kg0138, 3r,4r-1,3,4,5-tetrahydroxypentan-2-one, arabinulose, araboketose, erythropentulose
PubChem CID 151261
ChEBI CHEBI:17173
IUPAC Name (3R,4R)-1,3,4,5-tetrahydroxypentan-2-one
SMILES C(C(C(C(=O)CO)O)O)O

Specifications

Color Colorless
Quantity 25 mg
Formula Weight 150.1
Percent Purity 0.997
Physical Form Liquid
Chemical Name or Material D-ribulose, For HPLC analysis
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.