Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-Sorbitol, MP Biomedicals™

Catalog No. MP021029381 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP021029381 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP021029381 Supplier MP Biomedicals, Inc Supplier No. 02102938.1

Commonly used as a sugar substitute. D-Sorbitol, MP Biomedicals is a carbon sugar alcohol obtained by reduction of glucose, changing the aldehyde group to a hydroxyl group. It may be used for washing spheroplasts and in isoelectric focusing to minimize en

D-Sorbitol is a sugar alcohol that occurs naturally and is also produced synthetically from glucose. Naturally, D-sorbitol occurs widely in plants via photosynthesis, ranging from algae to higher order fruits of the family Rosaceae.

The food industry uses D-sorbitol as an additive in the form of a sweetener, humectant, emulsifier, thickener, or dietary supplement. D-Sorbitol has also been found in cosmetics, paper, and pharmaceuticals. It was formerly used as a diuretic and may still be used as a laxative and in irrigating solutions for some surgical procedures.

D-Sorbitol may be used for washing spheroplasts and in isoelectric focusing to minimize endoosmotic flow in agarose gels. It may also be used to induce osmotic stress. Sorbitol can be used as a screen for the O154:H7 strain of E. coli, since this strain is one of the few strains which cannot metabolize sorbitol.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-70-4
Molecular Formula C6H14O6
Molecular Weight (g/mol) 182.172
InChI Key FBPFZTCFMRRESA-JGWLITMVSA-N
Synonym d-sorbitol, sorbitol, d-glucitol, glucitol, l-gulitol, --sorbitol, diakarmon, glucarine, sorbol, d---sorbitol
PubChem CID 5780
ChEBI CHEBI:17924
IUPAC Name (2R,3R,4R,5S)-hexane-1,2,3,4,5,6-hexol
SMILES C(C(C(C(C(CO)O)O)O)O)O

Specifications

Melting Point 95°C to 105°C
Density 1.47g/mL at −5°C (Lit.)
Quantity 100 g
Solubility Information Soluble in water (20g/100mL); insoluble in alcohol.
Optical Rotation −2° ± 2° (c=2, water)
Formula Weight 182.17
Percent Purity ≥98% Anhydrous
Physical Form Crystalline Powder
Recommended Storage Store at Room Temperature (15–30°C), dessicated.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.