Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-Sorbitol (Powder/Laboratory), Fisher Chemical

Catalog No. s459500 Shop All Fisherbrand Products
Click to view available options
Quantity:
500 g

Chemical Identifiers

CAS 50-70-4
Molecular Formula C6H14O6
Molecular Weight (g/mol) 182.172
MDL Number MFCD00004708
InChI Key FBPFZTCFMRRESA-JGWLITMVSA-N
Synonym d-sorbitol, sorbitol, d-glucitol, glucitol, l-gulitol, --sorbitol, diakarmon, glucarine, sorbol, d---sorbitol
PubChem CID 5780
ChEBI CHEBI:17924
IUPAC Name (2R,3R,4R,5S)-hexane-1,2,3,4,5,6-hexol
SMILES C(C(C(C(C(CO)O)O)O)O)O

Specifications

Alpha Vector Sorbitol
Melting Point 75°C
Color White
Density 1.489g/cm³
pH 6 to 7
Boiling Point 295°C
Packaging Poly Bottle
Linear Formula CH2OH(CHOH)4CH2OH
Quantity 500 g
Formula Weight 182.17
Grade Laboratory
Physical Form Solid
Chemical Name or Material D-Sorbitol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.