Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(+)-Tubocurarine chloride pentahydrate, 98+%

Catalog No. aaj60222mc
Click to view available options
Quantity:
100 mg

Competitive, non-selective nicotinic acetylcholine receptor antagonist | CAS: 57-94-3 | C37H43Cl2N2O6+ | 682.659 g/mol

Competitive, non-selective nicotinic acetylcholine receptor antagonist

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 57-94-3
Melting Point 268°C (decomposition)
Color White
Molecular Formula C37H43Cl2N2O6+
Quantity 100 mg
MDL Number MFCD00013471
UN Number UN1544
Sensitivity Air sensitive
Merck Index 14,9807
Synonym tubocurarine chloride
Solubility Information Soluble in water to 50mg/ml, with warming as needed. Also soluble in ethanol or methanol
InChI Key GXFZCDMWGMFGFL-KKXMJGKMSA-O
SMILES CN1CCC2=CC(=C3C=C2C1CC4=CC=C(C=C4)OC5=C6C(CC7=CC(=C(C=C7)O)O3)[N+](CCC6=CC(=C5O)OC)(C)C)OC.Cl.Cl
Molecular Weight (g/mol) 682.659
PubChem CID 129894468
Formula Weight 681.65 (Anhydrous)
Physical Form Crystalline Powder
Chemical Name or Material D-Tubocurarine chloride
Show More Show Less
Hazard Category H301
Hazard Statement GHS H Statement
H300
Fatal if swallowed
Precautionary Statement P264b-P270-P301+P310-P330-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 615-023-7
RTECSNumber YO5100000
TSCA No
Recommended Storage Keep cold; Store under Argon

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.