Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-(-)-Valinol, 98%

Catalog No. aal1416603
Click to view available options
Quantity:
1 g
5 g

Catalog No. AAL1416603

Supplier: Thermo Scientific Chemicals L1416603

Only null left
Add to Cart

CAS: 4276-09-9 | C5H13NO | 103.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4276-09-9
Molecular Formula C5H13NO
Molecular Weight (g/mol) 103.17
MDL Number MFCD00064297
InChI Key NWYYWIJOWOLJNR-YFKPBYRVSA-N
Synonym d-valinol, r---2-amino-3-methyl-1-butanol, r-2-amino-3-methylbutan-1-ol, 2r-2-amino-3-methylbutan-1-ol, r-2-amino-3-methyl-1-butanol, d-2-amino-3-methyl-1-butanol, 2r-2-amino-3-methyl-1-butanol, 1r-1-isopropyl-2-hydroxyethylamine, d---valinol r---2-amino-3-methyl-1-butanol, d-valinol r---2-amino-3-methyl-1-butanol
PubChem CID 6950587
IUPAC Name (2R)-2-amino-3-methylbutan-1-ol
SMILES CC(C)[C@@H](N)CO

Specifications

Refractive Index 1.455
Density 0.931
Boiling Point 189°C to 190°C
Melting Point 30°C to 34°C
Quantity 1 g
Flash Point 78°C (172°F)
Optical Rotation −11° (c=10 in Water)
Sensitivity Air sensitive
Beilstein 1719138
Formula Weight 103.17
Percent Purity 98%
Chemical Name or Material D-(-)-Valinol
Show More Show Less
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.