Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D4-GDI/RhoGDI2 Antibody (10D774), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NB339550 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB339550 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB339550 Supplier Novus Biologicals Supplier No. NB10056556AF350
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

D4-GDI/RhoGDI2 Monoclonal specifically detects D4-GDI/RhoGDI2 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Western Blot.

Specifications

Antigen D4-GDI/RhoGDI2
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 10D774
Conjugate Alexa Fluor 350
Dilution Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias ARHGDIB Rho GDP dissociation inhibitor (GDI) beta, D4, GDIA2, GDID4, Ly-GDI, LYGDI, RAP1GN1, RhoGDI2
Host Species Mouse
Immunogen A synthetic peptide corresponding to human D4-GDI/RhoGDI2 was used as the immunogen.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 397
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.