Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ D609
GREENER_CHOICESDP

Catalog No. 143750 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-375-0 50 mg
14-371-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 14-375-0 Supplier Tocris Bioscience™ Supplier No. 1437/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective PC-PLC inhibitor

Selective competitive phosphatidyl choline-specific phospholipase C (PC-PLC) inhibitor (Ki = 6.4 μM); antiviral and antitumor agent. Also inhibits sphingomyelin synthase. Suppresses LPS- and IFNγ-induced NO production (IC50 = 20 mg/ml) and blocks oxidative glutamate toxicity in nerve cells. Antioxidant in vivo.

Chemical Identifiers

CAS 83373-60-8
Molecular Formula C11H18KOS2

Specifications

Quantity 50 mg
Formula Weight 266.46
Purity >98%
Molecular Weight (g/mol) 269.482
Chemical Name or Material D609
Recommended Storage Desiccate at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.