Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DAAM1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA512293
Change view
Click to view available options
Quantity:
400 μL
Catalog No. Quantity
PIPA512293 400 μL
1 options

Catalog No. PIPA512293

Supplier: Invitrogen™ PA512293

Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DAAM1 Polyclonal Antibody for Western Blot, IHC (P)

DAAM1 is a diaphanous-related formin first studied as a novel Disheveled binding protein and shown to be crucial for the planar cell polarity pathway in Xenopus. DAAM1, like other formins, directs nucleation and elongation of new actin filaments using its conserved formin-homology-2 (FH2) domain. It is a Rho-regulated formin implicated in neuronal development and regulation of gastrulation. It was identified as a binding protein of the Wnt receptor-associated protein Disheveled. It also binds the GTP-bound RhoA and increases its GTP-bound form population in cells. It exists in an autoinhibited state via intramolecular interactions between its amino-terminal GBD and carboxyl-terminal DAD domains. Structurally the protein includes a GTPase-binding domain (GBD), a diaphanous inhibitory domain (DID), and a coiled-coil region followed by the FH1, FH2 and C-terminal diaphanous auto-regulatory domain (DAD). In humans, the DAAM1 gene is mapped to chromosome 14q23.1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DAAM1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene DAAM1
Gene Accession No. Q9Y4D1
Gene Alias Daam1; Disheveled-associated activator of morphogenesis 1; dishevelled associated activator of morphogenesis 1; I79_021650; KIAA0666
Gene Symbols DAAM1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 1034-1063 amino acids from the C-terminal region of human DAAM1.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23002
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.