Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DAB2IP Antibody, Novus Biologicals™
SDP

Catalog No. NBP130081 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP130081 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP130081 Supplier Novus Biologicals Supplier No. NBP130081
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DAB2IP Polyclonal specifically detects DAB2IP in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen DAB2IP
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000-1:10000, Immunoprecipitation 2-5 ug/mg lysate
Formulation TBS and 0.1% BSA with 0.09% Sodium Azide
Gene Accession No. Q5VWQ8
Gene Alias AIP1FLJ39072, ASK-interacting protein 1, DAB2 interacting protein, DAB2 interaction protein, DAB2-interacting protein, DIP1/2, disabled homolog 2-interacting protein, DOC-2/DAB2 interactive protein, KIAA1743AF9Q34ASK1-interacting protein 1, nGAP-like protein
Gene Symbols DAB2IP
Host Species Rabbit
Immunogen Antigen maps to a region between residue 850 and 900 of human disabled homolog 2 interacting protein using the numbering given in entry CAQ10385.1
Molecular Weight of Antigen 132 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 153090
Test Specificity Based on 100% sequence identity, this antibody is predicted to react with Panda, Rhesus Monkey, Gorilla, Chimpanzee, White-tufted-ear marmoset, Naked mole rat, African elephant, Northern white-cheeked gibbon, and Thirteen-lined ground squirrel.
Target Species Human, Mouse
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.