Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DACH1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP227192 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP227192 0.1 mg
NBP227192SS 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP227192 Supplier Novus Biologicals Supplier No. NBP227192
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DACH1 Polyclonal specifically detects DACH1 in Bovine, Canine, Chicken, Equine, Human, Mouse, Primate samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot.

Specifications

Antigen DACH1
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 5-7ug/ml∼, Immunohistochemistry-Paraffin 5ug/ml
Formulation 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
Gene Alias Dach1, DACHdachshund homolog 1, dachshund homolog (Drosophila), dachshund homolog 1 (Drosophila), FLJ10138
Gene Symbols DACH1
Host Species Rabbit
Immunogen Amino acids 474-501 of human Dach1 were used as the immunogen.
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Neuronal Stem Cell Markers, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 1602
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human (isoform CRA_a: 676 amino acids, CRA_d: 708 amino acids, CRA_e: 417 amino acids), monkey, mouse, horse, dog, cow and chicken, 94% homologous in xenopus and 83% homologous in zebrafish. The immunogen is also 72% homologous in Dach2.
Target Species Bovine, Canine, Chicken, Equine, Human, Mouse, Primate
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.