Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DACH1 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$206.50 - $426.00

Specifications

Antigen DACH1
Concentration 0.5 mg/mL
Dilution Western Blot 5-7ug/ml∼, Immunohistochemistry-Paraffin 5ug/ml
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP227192SS
SDP
View Documents
Novus Biologicals
NBP227192SS
0.025 mg
Each for $206.50
Only null left
Add to Cart
 
NBP227192
SDP
View Documents
Novus Biologicals
NBP227192
0.1 mg
Each for $426.00
Only null left
Add to Cart
 
Description

Description

DACH1 Polyclonal specifically detects DACH1 in Human, Mouse, Bovine, Canine, Chicken, Equine, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

DACH1
Western Blot 5-7ug/ml∼, Immunohistochemistry-Paraffin 5ug/ml
Polyclonal
Purified
RUO
Human, Mouse, Bovine, Canine, Chicken, Equine, Primate
Dach1, DACHdachshund homolog 1, dachshund homolog (Drosophila), dachshund homolog 1 (Drosophila), FLJ10138
DACH1
IgG
Protein A purified
The amino acid sequence used as immunogen is 100% homologous in human (isoform CRA_a: 676 amino acids, CRA_d: 708 amino acids, CRA_e: 417 amino acids), monkey, mouse, horse, dog, cow and chicken, 94% homologous in xenopus and 83% homologous in zebrafish. The immunogen is also 72% homologous in Dach2.
0.5 mg/mL
Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Neuronal Stem Cell Markers, Stem Cell Markers
0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
1602
Amino acids 474-501 of human Dach1 were used as the immunogen.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.