Learn More
DACH1 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31
Specifications
Antigen | DACH1 |
---|---|
Applications | Immunohistochemistry, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-156-5462
![]() |
Bethyl Laboratories, Inc
A303556AT |
20 μL |
Each of 1 for $151.31
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections. Based on 100% sequence identity, this antibody is predict
Dachshund homolog 1 belongs to the DACH/Dachshund family. It is a transcription factor involved in the regulation of organogenesis. DACH1 plays a role in retinal and pituitary precursor cell proliferation.It is reported to be expressed in breast cancer cell lines and to inhibit transforming growth factor-beta (TGF-beta)-induced apoptosis. DACH1 repressed TGF-beta induction of AP-1 and Smad signaling in gene reporter assays and repressed endogenous TGF-beta-responsive genes by microarray analyses. DACH1 participates in the negative regulation of TGF-beta signaling by interacting with NCoR and Smad4. Mouse DACH1 protein was immunolocalized to specific cell types within the developing kidneys, eyes, cochleae, and limb buds.Specifications
DACH1 | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
OTTHUMP00000040839; dachshund homolog 1 (Drosophila) ; DACH1; RP11-512J14.1; DACH; FLJ10138; dachshund homolog 1 | |
DACH1 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
Immunohistochemistry, Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q9UI36 | |
1602 | |
Between 740 and 760 | |
Primary | |
4° C | |
DACH1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.