Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Dacomitinib
GREENER_CHOICE

Catalog No. p-7114681
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent irreversible pan ErbB inhibitor

Dacomitinib is a potent irreversible pan ErbB inhibitor (IC50 values are 6.0, 45.7 and 73.7 nM for EGFR, ERBB2 and ERBB4, respectively). Inhibits mutant and wild type forms of ErbB, including EGFR T780M, and shows activity in NSCLC cell lines sensitive and resistant to Iressa (Cat. No. 3000). Inhibits tumor growth. Orally bioavailable.

Specifications

Chemical Name or Material (2E)-N-[4-[(3-Chloro-4-fluorophenyl)amino]-7-methoxy-6-quinazolinyl]-4-(1-piperidinyl)-2-butenamide
CAS 1110813-31-4
Assay Percent Range >98%
Quantity 10 mg
Molecular Formula C24H25ClFN5O2
Formula Weight Observed MW: 469.94
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.