Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dact1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP224768SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP224768SS 0.025 mg
NBP224768 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP224768SS Supplier Novus Biologicals Supplier No. NBP224768SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 1 publication

Dact1 Polyclonal specifically detects Dact1 in Chicken, Human, Mouse, Primate, Rat, Xenopus species, Zebrafish samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot.

Specifications

Antigen Dact1
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5 - 1 ug/mL, Immunohistochemistry-Paraffin 10 ug/mL
Formulation PBS, 0.05% BSA with 0.05% Sodium Azide
Gene Alias DAPPER, dapper, antagonist of beta-catenin, homolog 1 (Xenopus laevis), DAPPER1dapper homolog 1, antagonist of beta-catenin (xenopus), DPR1dapper homolog 1, antagonist of beta-catenin, FRODOdapper homolog 1, HDPR1, Hepatocellular carcinoma novel gene 3 protein, heptacellular carcinoma novel gene 3, HNG3, THYEX3
Gene Symbols DACT1
Host Species Rabbit
Immunogen A portion of amino acids 725-775 of human Dapper homolog 1 was used as the immunogen for the antibody.
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 51339
Test Specificity Based on immunogon sequence homology (76%), the antibody may cross-react with Dact2
Target Species Chicken, Human, Mouse, Primate, Rat, Xenopus species, Zebrafish
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.