Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DAF-3 Antibody, Novus Biologicals™
SDP

Catalog No. NB1001924SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1001924SS 0.025 mL
NB1001924 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB1001924SS Supplier Novus Biologicals Supplier No. NB1001924SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 3 publications

DAF-3 Polyclonal specifically detects DAF-3 in C. elegans samples. It is validated for Chemotaxis, ChIP Assay, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.

Specifications

Antigen DAF-3
Applications Chemotaxis, ChIP Assay, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:5000, Chromatin Immunoprecipitation reported in scientific literature (PMID 16777607), Immunohistochemistry 1:50-1:200, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:50-1:200, Chromatin Immunoprecipitation (ChIP) 1:10-1:500
Formulation Whole antisera with 0.05% Sodium Azide
Gene Accession No. O17532
Gene Alias Abnormal dauer formation protein 3, DAF 3, DAF3, isoform c
Host Species Rabbit
Immunogen A synthetic peptide made to the C-terminus (within residues 700-796) of the C.elegans DAF-3 protein. [UniProt# O17532]
Purification Method Unpurified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Chromatin Research, DNA Repair, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 180431
Test Specificity C. elegans.
Target Species C. elegans
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.