Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Daidzein, 98.7%, MP Biomedicals

Catalog No. icn15881280 Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
100 mg

Catalog No. ICN15881280

Supplier: MP Biomedicals, Inc 0215881280

Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 486-66-8
Molecular Formula C15H10O4
Molecular Weight (g/mol) 254.241
InChI Key ZQSIJRDFPHDXIC-UHFFFAOYSA-N
Synonym daidzein, 4',7-dihydroxyisoflavone, daidzeol, 7,4'-dihydroxyisoflavone, 7-hydroxy-3-4-hydroxyphenyl-4h-chromen-4-one, diadzein, 7-hydroxy-3-4-hydroxyphenyl-4h-1-benzopyran-4-one, 4h-1-benzopyran-4-one, 7-hydroxy-3-4-hydroxyphenyl, 7-hydroxy-3-4-hydroxyphenyl chromen-4-one, 7-hydroxy-3-4-hydroxyphenyl-4-benzopyrone
PubChem CID 5281708
ChEBI CHEBI:28197
IUPAC Name 7-hydroxy-3-(4-hydroxyphenyl)chromen-4-one
SMILES C1=CC(=CC=C1C2=COC3=C(C2=O)C=CC(=C3)O)O

Specifications

Color White
Assay Percent Range 0.987
Quantity 100 mg
Formula Weight 254.23
Physical Form Powder
Chemical Name or Material Daidzein, For HPLC analysis
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.