Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dansyl-L-alanine cyclohexylammonium salt, MP Biomedicals™

Catalog No. icn10000701 Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
1 g

Catalog No. ICN10000701

Supplier: MP Biomedicals, Inc 0210000701

Chemical Identifiers

CAS 53332-27-7
Molecular Formula C21H31N3O4S
Molecular Weight (g/mol) 421.556
InChI Key CZUBQNQWSOLSJH-PPHPATTJSA-N
Synonym dansyl-l-alanine cyclohexylammonium salt, n-5-dimethylamino-1-naphthyl sulphonyl-l-alanine, compound with cyclohexylamine 1:1, dansyl-l-alanine cyclohexylammonium salt, fluorescent amino acid, 2s-2-5-dimethylamino naphthalene-1-sulfonamido propanoic acid; cyclohexylamine, cyclohexanamine s-2-5-dimethylamino naphthalene-1-sulfonamido propanoate, cyclohexanamine; 2s-2-5-dimethylamino naphthalen-1-yl sulfonylamino propanoic acid, n-5-dimethylamino-1-naphthalenyl sulfonyl-l-alanine cyclohexylammonium salt
PubChem CID 6452802
IUPAC Name cyclohexanamine;(2S)-2-[[5-(dimethylamino)naphthalen-1-yl]sulfonylamino]propanoic acid
SMILES CC(C(=O)O)NS(=O)(=O)C1=CC=CC2=C1C=CC=C2N(C)C.C1CCC(CC1)N

Specifications

Color White
Quantity 1 g
Formula Weight 421.6
Physical Form Powder
Chemical Name or Material Dansyl-L-alanine cyclohexylammonium salt
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.