Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ DAO Human Recombinant Protein
SDP

Catalog No. NBP25223705 Shop All R&D Systems Products
Click to view available options
Quantity:
0.01 mg
0.05 mg

Highly purified and high bioactivity. Generating reliable and reproducible results. Applications: Functional, SDS-Page, Bioactivity

A bioactive recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-347 of Human DAO The Recombinant Human DAO Protein is derived from E. coli. The Recombinant Human DAO Protein has been validated for the following applications: Functional, SDS-Page, Bioactivity.

Specifications

Concentration 0.5mg/mL
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0) containing 20% glycerol 1mM DTT
Gene ID (Entrez) 1610
Molecular Weight (g/mol) TMW: 41.6kDa
Quantity 0.05 mg
Storage Requirements Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
Gene Alias DAAO, D-amino-acid oxidase, DAMOXOXDA, DAO, EC 1.4.3, EC 1.4.3.3, MGC35381
Common Name DAO
Biological Activity Specific activity is > 3.5 units/mg, which one unit will oxidatively deaminate 1.0 umole of D-alanine to pyruvate per minute at pH 8.5 at 37C, the presence of catalase.
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >90%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain
Show More Show Less

For research use only and is not approved for use in humans or in clinical diagnosis

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.