Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DAP Kinase 2 Antibody (OTI1C5), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. N271753A700 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N271753A700 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N271753A700 Supplier Novus Biologicals Supplier No. NBP271753AF700
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

DAP Kinase 2 Monoclonal specifically detects DAP Kinase 2 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen DAP Kinase 2
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI1C5
Conjugate Alexa Fluor 700
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias DAP kinase 2, DAP-kinase-related protein 1, DAP-kinase-related protein 1 beta isoform, death-associated protein kinase 2, DRP1, DRP-1MGC119312, EC 2.7.11, EC 2.7.11.1
Gene Symbols DAPK2
Host Species Mouse
Immunogen Full length recombinant protein of human DAP Kinase 2 (NP_055141) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Apoptosis, Autophagy, Cellular Markers
Primary or Secondary Primary
Gene ID (Entrez) 23604.0
Target Species Human, Mouse, Rat, Canine, Monkey
Content And Storage Store at 4°C in the dark.
Isotype IgG3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.