Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DAP10/HCST Antibody (982107), Alexa Fluor™ 700, R&D Systems™
SDP

Catalog No. NB003337 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB003337 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB003337 Supplier Novus Biologicals Supplier No. MAB97862AF700
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

DAP10/HCST Monoclonal specifically detects DAP10/HCST in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin).

Specifications

Antigen DAP10/HCST
Applications Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 982107
Conjugate Alexa Fluor 700
Dilution Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias DAP10PIK3AP, DKFZP586C1522, hematopoietic cell signal transducer, KAP10DNAX-activation protein 10, kinase assoc pro of ∽10kDa, kinase assoc protein, Membrane protein DAP10, phosphoinositide-3-kinase adaptor protein, Transmembrane adapter protein KAP10
Gene Symbols HCST
Host Species Mouse
Immunogen Synthetic peptide containing human DAP10/HCST
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immune System Diseases, Immunology, Innate Immunity
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.