Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DAP3 Antibody (42C617.1.2), Biotin, Novus Biologicals™
SDP

Catalog No. NB10056539B Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056539B 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10056539B Supplier Novus Biologicals Supplier No. NB10056539B
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

DAP3 Monoclonal specifically detects DAP3 in Human, Mouse samples. It is validated for Western Blot.

Specifications

Antigen DAP3
Applications Western Blot
Classification Monoclonal
Clone 42C617.1.2
Conjugate Biotin
Dilution Western Blot
Gene Accession No. P51398
Gene Alias bMRP-10, DAP-3Death-associated protein 3, death associated protein 3, DKFZp686G12159,28S ribosomal protein S29, mitochondrial, FLJ12817, MGC126058, MGC126059, mitochondrial 28S ribosomal protein S29, MRP-S29Ionizing radiation resistance conferring protein, MRPS29S29mt
Gene Symbols DAP3
Host Species Mouse
Immunogen The antibody was raised against a synthetic peptide corresponding to amino acids 51-66 of human DAP-3 protein.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 7818
Test Specificity In Jurkat, a 45 kDa band is observed.
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.