Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DARC Antibody (2C3) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB001499 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB001499 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB001499 Supplier Novus Biologicals Supplier No. NBP2751960.025MG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

DARC Monoclonal specifically detects DARC in Human samples. It is validated for Western Blot, ELISA, Flow (Intracellular), Immunofluorescence, Block/Neutralize.

Specifications

Antigen DARC
Applications Western Blot, ELISA, Flow Cytometry, Immunofluorescence, Blocking Assay, Neutralization
Classification Monoclonal
Clone 2C3
Conjugate Unconjugated
Dilution Western Blot, ELISA 1:100 - 1:2000, Flow (Intracellular) 1:10 - 1:1000, Immunofluorescence, Block/Neutralize
Formulation PBS
Gene Alias CCBP1, CD234, CD234 antigen, Dfy, Duffy antigen/chemokine receptor, Duffy blood group antigen, Duffy blood group, chemokine receptor, Fy glycoprotein, FYDuffy blood group, Glycoprotein D, GPDWBCQ1, GpFy, Plasmodium vivax receptor
Gene Symbols ACKR1
Host Species Mouse
Immunogen CHO cells transfected with cDNA encoding the Duffy glycoprotein.
Quantity 0.025 mg
Gene ID (Entrez) 2532
Test Specificity The antibody binds specifcally to the pentapeptide FEDVW of the Duffy receptor.
Target Species Human
Content And Storage Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.