Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DARC Antibody, DyLight 488, Novus Biologicals™
SDP

Catalog No. NB218186 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB218186 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB218186 Supplier Novus Biologicals Supplier No. NBP177278G
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DARC Polyclonal antibody specifically detects DARC in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen DARC
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 488
Formulation 50mM Sodium Borate
Gene Alias CCBP1, CD234, CD234 antigen, Dfy, Duffy antigen/chemokine receptor, Duffy blood group antigen, Duffy blood group, chemokine receptor, Fy glycoprotein, FYDuffy blood group, Glycoprotein D, GPDWBCQ1, GpFy, Plasmodium vivax receptor
Host Species Rabbit
Immunogen Antibody was raised against a 16 amino acid synthetic peptide from near the carboxy terminus of human DARC. The immunogen is located within the last 50 amino acids of DARC. Amino Acid Squence: LPEGWSSHLDTLGSKS
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 2532
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.