Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DARC Antibody, Novus Biologicals™
SDP

Catalog No. NBH24210025 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBH24210025 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBH24210025 Supplier Novus Biologicals Supplier No. NB10024210.025MG
Add to Cart
Add to Cart

Goat Polyclonal Antibody has been used in 7 publications

DARC Polyclonal specifically detects DARC in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Peptide ELISA, Western Blot.

Specifications

Antigen DARC
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Peptide ELISA, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.3-1 ug/ml, Flow Cytometry 1:1000, Immunohistochemistry 3 ug/ml, Immunohistochemistry-Paraffin 3 ug/ml, Peptide ELISA detection limit 1:16000
Formulation 0.5 mg/ml Tris (pH 7.3) and 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. Q16570
Gene Alias CCBP1, CD234, CD234 antigen, Dfy, Duffy antigen/chemokine receptor, Duffy blood group antigen, Duffy blood group, chemokine receptor, Fy glycoprotein, FYDuffy blood group, Glycoprotein D, GPDWBCQ1, GpFy, Plasmodium vivax receptor
Gene Symbols DARC
Host Species Goat
Immunogen Peptide with sequence HRAELSPSTENSSQLDFED-C corresponding to N-Terminus according to NP_001116423.1, NP_002027.2.
Molecular Weight of Antigen 36 kDa
Purification Method Affinity Purified
Quantity 0.025 mg
Research Discipline Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 2532
Test Specificity This antibody is expected to recognize both reported isoforms (NP_001116423.1; NP_002027.2).
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.