Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DARPP-32, p Thr75 Antibody, Novus Biologicals™
SDP

Catalog No. NB300234 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB300234 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB300234 Supplier Novus Biologicals Supplier No. NB300234
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 9 publications

DARPP-32 Polyclonal specifically detects DARPP-32 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen DARPP-32 (p Thr75)
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000
Formulation 10mM HEPES (pH 7.5), 0.15M NaCl, 0.1 mg/ml BSA and 50% Glycerol with No Preservative
Gene Accession No. Q6J4I0
Gene Alias protein phosphatase 1, regulatory (inhibitor) subunit 1B, regulatory (inhibitor) subunit 1B (dopamine and cAMPregulated phosphoprotein, DARPP-32)
Gene Symbols PPP1R1B
Host Species Rabbit
Immunogen Synthetic phospho-peptide corresponding to amino acid residues surrounding Thr75 conjugated to KLH. Accession # Q6J4I0
Molecular Weight of Antigen 32 kDa
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Neuronal Cell Markers, Neuroscience, Neurotransmission, Phospho Specific, Protein Phosphatase, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 84152
Test Specificity Specific for the ∼32k DARPP-32 protein phosphorylated at Thr75. Immunolabeling is blocked by lambda-phosphatase treatment.
Target Species Human, Mouse, Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.