Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DARS Antibody, mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB329157 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB329157 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB329157 Supplier Novus Biologicals Supplier No. NBP300268MFV610
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DARS Polyclonal antibody specifically detects DARS in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation, Immunohistochemistry (Paraffin)

Specifications

Antigen DARS
Applications Western Blot, ELISA, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Dilution Western Blot, ELISA, Immunoprecipitation, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias aspartate tRNA ligase 1, cytoplasmic, Aspartate--tRNA ligase, aspartyl-tRNA synthetase, aspartyl-tRNA synthetase, cytoplasmic, aspRS, Cell proliferation-inducing gene 40 protein, cell proliferation-inducing protein 40, DKFZp781B11202, EC 6.1.1.12, MGC111579
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Human DARS (Accession#: P14868; Met1-Pro501)
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 1615
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.