Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ DAU 5884 hydrochloride

Click to view available options
Quantity:
10 mg
50 mg
Description
Selective muscarinic M3 receptor antagonist. In bovine tracheal smooth muscle, inhibits methacholine-dependent effects on cell proliferation and muscle contractility.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 351.83 |
| Purity | >99% |
| Molecular Weight (g/mol) | 351.831 |
| Chemical Name or Material | DAU 5884 hydrochloride |
Safety and Handling
| Recommended Storage | Desiccate at +4°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction