Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Daunomycin hydrochloride, MP Biomedicals™

Catalog No. icn15077701 Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
1 mg
5 mg
50 mg

Catalog No. ICN15077701

Supplier: MP Biomedicals, Inc 0215077701

Only null left
Add to Cart
Add to Cart
Photostability, Antileukemic, and drug metabolism studies
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23541-50-6
Synonym Daunorubicin hydrochloride, Leukaemomycin C

Specifications

Type Daunomycin Hydrochloride
Physical Form Solid
Quantity 1 mg
Molecular Formula C27H30ClNO10
MDL Number MFCD04974507
InChI Key GUGHGUXZJWAIAS-QQYBVWGSSA-N
SMILES CC1C(C(CC(O1)OC2CC(CC3=C(C4=C(C(=C23)O)C(=O)C5=C(C4=O)C=CC=C5OC)O)(C(=O)C)O)N)O.Cl
IUPAC Name (7S,9S)-9-acetyl-7-[(2R,4S,5S,6S)-4-amino-5-hydroxy-6-methyloxan-2-yl]oxy-6,9,11-trihydroxy-4-methoxy-8,10-dihydro-7H-tetracene-5,12-dione;hydrochloride
Molecular Weight (g/mol) 563.984
PubChem CID 62770
Formula Weight 563.99
Chemical Name or Material Daunomycin hydrochloride
Show More Show Less
storageNote1 Protect from light.
Recommended Storage Store at 4°C.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.