Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Daunorubicin HydroChloride, PROMO, Thermo Scientific Chemicals

Catalog No. J60224MAPM
Click to view available options
Quantity:
10 mg

Chemical Identifiers

CAS 23541-50-6
Molecular Formula C27H30ClNO10
Molecular Weight (g/mol) 563.98 g/mol
MDL Number MFCD04974507
InChI Key GUGHGUXZJWAIAS-QQYBVWGSSA-N
Synonym daunorubicin hydrochloride, daunorubicin hcl, daunomycin hydrochloride, daunoblastina, rubidomycin hydrochloride, daunorubicinol hydrochloride, daunoblastin, rubomycin, ondena, daunomycin chlorohydrate
PubChem CID 62770
IUPAC Name (7S,9S)-9-acetyl-7-[(2R,4S,5S,6S)-4-amino-5-hydroxy-6-methyloxan-2-yl]oxy-6,9,11-trihydroxy-4-methoxy-8,10-dihydro-7H-tetracene-5,12-dione;hydrochloride
SMILES CC1C(C(CC(O1)OC2CC(CC3=C(C4=C(C(=C23)O)C(=O)C5=C(C4=O)C=CC=C5OC)O)(C(=O)C)O)N)O.Cl

Specifications

Color Red
Quantity 10 mg
UN Number UN2811
Beilstein 4229221
Sensitivity Hygroscopic
Merck Index 14,2832
Physical Form Powder
Chemical Name or Material Daunorubicin hydrochloride
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 245-723-4
RTECSNumber HB7878000
TSCA No
Recommended Storage Keep cold

RUO - Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.