Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DAX1/NR0B1 Antibody (OTI5F5), DyLight 405, Novus Biologicals™
SDP

Catalog No. NBP270116V Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP270116V 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP270116V Supplier Novus Biologicals Supplier No. NBP270116V
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

DAX1/NR0B1 Monoclonal specifically detects DAX1/NR0B1 in Human samples. It is validated for Immunohistochemistry.

Specifications

Antigen DAX1/NR0B1
Applications Immunohistochemistry
Classification Monoclonal
Clone OTI5F5
Conjugate DyLight 405
Dilution Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias AHCDSS, AHCH, AHX, DAX1DAX-1, dosage-sensitive sex reversal, DSS-AHC critical region on the X chromosome protein 1, GTD, HHG, NROB1, nuclear hormone receptor, Nuclear receptor DAX-1, nuclear receptor subfamily 0 group B member 1, nuclear receptor subfamily 0, group B, member 1
Gene Symbols NR0B1
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 161-430 of human DAX1/NR0B1 (NP_000466) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Cancer, GPCR
Primary or Secondary Primary
Gene ID (Entrez) 190.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.