Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DAX1/NR0B1 Rabbit anti-Human, Clone: JU33-05, Novus Biologicals™
SDP

Catalog No. NBP275462 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP275462 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP275462 Supplier Novus Biologicals Supplier No. NBP275462

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Rabbit Monoclonal Antibody

DAX1/NR0B1 Monoclonal specifically detects DAX1/NR0B1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen DAX1/NR0B1
Applications Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone JU33-05
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, Immunocytochemistry/Immunofluorescence 1:10 - 1:500
Formulation TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide
Gene Alias AHCDSS, AHCH, AHX, DAX1DAX-1, dosage-sensitive sex reversal, DSS-AHC critical region on the X chromosome protein 1, GTD, HHG, NROB1, nuclear hormone receptor, Nuclear receptor DAX-1, nuclear receptor subfamily 0 group B member 1, nuclear receptor subfamily 0, group B, member 1
Gene Symbols NR0B1
Host Species Rabbit
Immunogen Synthetic peptide within Human DAX1/NR0B1 aa 1-50/ 470. (SwissProt: P51843 Human)
Molecular Weight of Antigen 52 kDa
Purification Method Protein A purified
Quantity 100 μL
Research Discipline Cancer, GPCR
Primary or Secondary Primary
Gene ID (Entrez) 190.0
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.