Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Daxx Antibody (DAXX-01), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NB50491A750 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB50491A750 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB50491A750 Supplier Novus Biologicals Supplier No. NB500491AF750
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Daxx Monoclonal specifically detects Daxx in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Daxx
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone DAXX-01
Conjugate Alexa Fluor 750
Dilution Western Blot, Immunocytochemistry/Immunofluorescence
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias BING2, CENP-C binding protein, DAP6MGC126245, Daxx, death domain-associated protein 6, death-associated protein 6, death-domain associated protein, EAP1, ETS1-associated protein 1, Fas death domain-associated protein, Fas-binding protein, hDaxx, MGC126246
Gene Symbols DAXX
Host Species Mouse
Immunogen Recombinant C-terminal part (aa 558-740) of human Daxx.
Purification Method Protein A purified
Quantity 0.1 mL
Research Discipline Apoptosis, Cancer, Phospho Specific, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 1616.0
Test Specificity The antibody DAXX-01 reacts with Daxx, a death domain containing protein mainly expressed in fetal and adult human and mouse tissue.
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.