Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Daxx Antibody (PCRP-DAXX-8B7), DyLight 488, Novus Biologicals™
SDP

Catalog No. NB384761 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB384761 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB384761 Supplier Novus Biologicals Supplier No. NBP308740G
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Daxx Monoclonal antibody specifically detects Daxx in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Protein Array

Specifications

Antigen Daxx
Applications Flow Cytometry, Immunofluorescence, Protein Array
Classification Monoclonal
Clone PCRP-DAXX-8B7
Conjugate DyLight 488
Formulation 50mM Sodium Borate
Gene Alias BING2, CENP-C binding protein, DAP6MGC126245, Daxx, death domain-associated protein 6, death-associated protein 6, death-domain associated protein, EAP1, ETS1-associated protein 1, Fas death domain-associated protein, Fas-binding protein, hDaxx, MGC126246
Host Species Mouse
Immunogen Recombinant full-length human Daxx protein (Uniprot: Q9UER7)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 1616
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.