Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DAZ4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. p-7206281 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-9184 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-9184 Supplier Proteintech Group Inc Supplier No. 140651AP150UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene is a member of the DAZ gene family and is a candidate for the human Y-chromosomal azoospermia factor (AZF). This RNA-binding protein is important for spermatogenesis.

Specifications

Antigen DAZ4
Applications Western Blot
Classification Polyclonal
Concentration 0.19 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene DAZ4
Gene Accession No. Q86SG3
Gene Alias DAZ, DAZ1, DAZ4, deleted in azoospermia 4, pDP1680, pDP1681
Gene Symbols DAZ4
Host Species Rabbit
Immunogen DAZ4 Fusion Protein Ag5116
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57135
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.