Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DAZL Antibody (DAZL/4253), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NB384752 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB384752 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB384752 Supplier Novus Biologicals Supplier No. NBP308742AF594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

DAZL Monoclonal antibody specifically detects DAZL in Human, Mouse, Rat, Cynomolgus Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen DAZL
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone DAZL/4253
Conjugate Alexa Fluor 594
Formulation 50mM Sodium Borate
Gene Alias DAZHSPGY-like-autosomal, DAZL1DAZ-like autosomal, DAZLA, deleted in azoospermia-like, Deleted in azoospermia-like 1, germline specific RNA binding protein, MGC26406, spermatogenesis gene on the Y-like autosomal, SPGYLADAZ homolog
Host Species Mouse
Immunogen Recognizes DAZL protein C terminus sequence CRVHHFRRSRAMLKSV. (Uniprot: Q92904)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1618
Target Species Human, Mouse, Rat, Cynomolgus Monkey
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.