Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DAZL Antibody (DAZL3/11A) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NBP261937 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP261937 0.1 mg
NB435231 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP261937 Supplier Novus Biologicals Supplier No. NBP261937

Mouse Monoclonal Antibody

DAZL Monoclonal specifically detects DAZL in Human, Mouse, Rat, Cynomolgus Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen DAZL
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone DAZL3/11A
Concentration 1mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:1000, Immunohistochemistry 1:50 - 1:500, Immunocytochemistry/Immunofluorescence 1:50 - 1:500
Formulation PBS with 0.02% Sodium Azide
Gene Alias DAZHSPGY-like-autosomal, DAZL1DAZ-like autosomal, DAZLA, deleted in azoospermia-like, Deleted in azoospermia-like 1, germline specific RNA binding protein, MGC26406, spermatogenesis gene on the Y-like autosomal, SPGYLADAZ homolog
Gene Symbols DAZL
Host Species Mouse
Immunogen Recognizes DAZL protein C terminus sequence CRVHHFRRSRAMLKSV
Purification Method Protein A purified
Quantity 0.1 mg
Primary or Secondary Primary
Gene ID (Entrez) 1618
Target Species Cynomolgus, Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.