Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DAZL Antibody (DAZL3/11A) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$196.10
Specifications
Antigen | DAZL |
---|---|
Clone | DAZL3/11A |
Dilution | Western Blot 1:100-1:1000, Immunohistochemistry 1:50 - 1:500, Immunocytochemistry/Immunofluorescence 1:50 - 1:500 |
Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot |
Classification | Monoclonal |
Description
DAZL Monoclonal specifically detects DAZL in Human, Mouse, Rat, Cynomolgus Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
DAZL | |
Western Blot 1:100-1:1000, Immunohistochemistry 1:50 - 1:500, Immunocytochemistry/Immunofluorescence 1:50 - 1:500 | |
Monoclonal | |
Purified | |
Cynomolgus, Human, Mouse, Rat | |
DAZHSPGY-like-autosomal, DAZL1DAZ-like autosomal, DAZLA, deleted in azoospermia-like, Deleted in azoospermia-like 1, germline specific RNA binding protein, MGC26406, spermatogenesis gene on the Y-like autosomal, SPGYLADAZ homolog | |
DAZL | |
IgG1 | |
Protein A purified |
DAZL3/11A | |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot | |
Unconjugated | |
Mouse | |
PBS with 0.02% Sodium Azide | |
1618 | |
Recognizes DAZL protein C terminus sequence CRVHHFRRSRAMLKSV | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title