Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DBF4 Antibody, Novus Biologicals™
SDP

Catalog No. NBP178282 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP178282 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP178282 Supplier Novus Biologicals Supplier No. NBP178282
Add to Cart
Add to Cart

Goat Polyclonal Antibody

DBF4 Polyclonal specifically detects DBF4 in Mouse, Rat samples. It is validated for Western Blot, Peptide ELISA.

Specifications

Antigen DBF4
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1 - 3 ug/ml, Peptide ELISA Detection limit 1:16000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Alias ASKchif, DBF4 homolog (S. cerevisiae), DBF4ACHIF, DBF4-type zinc finger-containing protein 1, ZDBF1DBF-type containing 1
Gene Symbols DBF4
Host Species Goat
Immunogen Peptide with sequence C-DKKRTEYLPAHEDRT corresponding to internal region according to NP_001177646.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 10926
Test Specificity This antibody is expected to recognize both reported isoforms (NP_038754.1; NP_001177646.1).
Target Species Mouse, Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.