Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DBF4B Antibody, Novus Biologicals™
SDP

Catalog No. NB301675 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB301675 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB301675 Supplier Novus Biologicals Supplier No. NBP324777
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DBF4B Polyclonal antibody specifically detects DBF4B in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen DBF4B
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500
Formulation PBS, pH 7.2, 40% glycerol
Gene Alias Activator of S phase kinase-like protein 1, activator of S-phase kinase-like protein 1, ASKL1CHIFB, ASK-like protein 1, chifb, Chiffon homolog B, DBF4 homolog B (S. cerevisiae), Dbf4-related factor 1, DRF1MGC15009, FLJ13087, protein DBF4 homolog B, ZDBF1B, zinc finger, DBF-type containing 1B
Host Species Rabbit
Immunogen This antibody has been engineered to specifically recognize the recombinant protein DBF4B using the following amino acid sequence: GKGDDCLELESSMAESRLRAPDLGVSRCLGKCQKNSPGARKHPFSGKSFYLDLPAGKNLQFLTGAIQQLGGVIEGFLSKEVSYIVSSRREVKAESSGKSHRGCPSPSPSEVRVETSAMVDPKGSHPRPSRKPVDSV
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80174
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.