Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ DBI-SALA™ Confined Space Vehicle Hitch Mount Sleeve Davit Base

3M™ DBI-SALA™ Confined Space Vehicle Hitch Mount Sleeve Davit Base features a stabilizer T-bar.

$3498.89 - $5311.43

Specifications

Color Green
Product Type Vehicle Hitch Mount Sleeve Davit Base
View More Specs

Products 2
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity & Availability  
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity & Availability  
19-809-717
View Documents
3M™
7100314312
27.5 in. (69.8 cm) maximum offset davit mast
Case for $5,311.43
Only null left
Add to Cart
 
19-809-729
View Documents
3M™
7012820655
42.5 in. (108 cm) maximum offset davit mast
Case for $3,498.89
Only null left
Add to Cart
 
Description

Description

  • Vehicle hitch mount sleeve davit base
  • Installs into a 2 in. (5.1 cm) ball hitch
  • Adjustable stabilizer T-bar
  • Detent style height adjustment
  • PVC davit mast sleeve liner
  • Set-up and dismantle without tools
  • Durable and corrosion resistant construction
Specifications

Specifications

Green
Vehicle Hitch Mount Sleeve Davit Base
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.