Learn More
3M™ DBI-SALA™ ExoFit™ X300 Comfort Vest Climbing Safety Harness
Catalog No. 13051055 Shop All 3M Products
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
Buckle Style (Leg):
Auto-Locking Quick-Connect
Tongue Buckle
Length (English):
15.9 in.
16.0 in.
17.0 in.
18.75 in.
20.0 in.
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Buckle Style (Leg) Length (English)
13-051-055 X-Small Auto-Locking Quick-Connect 18.75 in.
13-051-004 Small Auto-Locking Quick-Connect 16.0 in.
13-051-110 Small Tongue Buckle 16.0 in.
13-051-005 Medium Auto-Locking Quick-Connect 17.0 in.
13-051-111 Medium Tongue Buckle 16.0 in.
13-051-006 Large Auto-Locking Quick-Connect 20.0 in.
13-051-112 Large Tongue Buckle 16.0 in.
13-051-007 X-Large Auto-Locking Quick-Connect 15.9 in.
13-051-113 X-Large Tongue Buckle 16.0 in.
13-051-008 2X-Large Auto-Locking Quick-Connect 18.75 in.
13-051-114 2X-Large Tongue Buckle 16.0 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
This item is not returnable. View return policy
For industrial/occupational use only. Not for consumer sale or use.
Catalog No. 13-051-055 Supplier 3M™ Supplier No. 7012816126
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
For industrial/occupational use only. Not for consumer sale or use.
The 3M™ DBI-SALA™ ExoFit™ X300 is designed for those who refuse to compromise. The X300 harnesses are outfitted with high-quality components that have been developed to enhance the safety, convenience and performance for working at height on your job site.
  • The X300 comfort harness is built with 6000 lb. polyester webbing, robust nylon/polyester hybrid comfort padding with reflective 3M Scotchlite™ panels, and lightweight aluminum hardware
  • Revolving Torso Adapters adjust harness fit with ease and speed and lock webbing into place to maintain a comfortable and safe harness fit
  • Equipped with anti-sliding dorsal D-ring pad to help greatly reduce dorsal D-ring slide
  • Integrated easy-link SRL adapter lets you quickly and easily connect and disconnect personal SRLs
  • Auto-locking quick connect buckles secure contact with webbing, reducing the need to readjust
  • Automatic stand-up D-ring (lightweight aluminum) enables efficient dorsal connections
  • Low-profile suspension trauma strap design integrated between harness webbing
  • Equipped with auto-resetting lanyard keepers for fast and easy parking of snap hooks and carabiners
  • 3M™ Connected Safety ID (RFID) integrated for simplified inventory, inspection, and record management
TRUSTED_SUSTAINABILITY
Specifications
Color Gray
Material Polyester
Length (English) 18.75 in.
Length (Metric) 47.625 cm
Buckle Style (Chest) Auto-Locking Quick-Connect
Buckle Style (Leg) Auto-Locking Quick-Connect
D-Ring Location Back, Front
Product Type Climbing Safety Harness
Size X-Small
3M industrial and occupational products are intended, labeled, and packaged for sale to trained industrial and occupational customers for workplace use. Unless specifically stated otherwise on the applicable product packaging or literature, these products are not intended, labeled, or packaged for sale to or use by consumers (e.g., for home, personal, primary or secondary school, recreational/sporting, or other uses not described in the applicable product packaging or literature), and must be selected and used in compliance with applicable health and safety regulations and standards (e.g., U.S. OSHA, ANSI), as well as all product literature, user instructions, warnings, and other limitations, and the user must take any action required under any recall, field action, or other product use notice. Misuse of 3M industrial and occupational products may result in injury, sickness, death, or property damage. For help with product selection and use, consult your on-site safety professional, industrial hygienist, or other subject matter expert.