Learn More
3M™ DBI-Sala™ Hi-Vis WorkVest and Harness Combination
Shop All 3M Products
Click to view available options
Size:
Large Vest; Universal Harness
Medium Vest; Universal Harness
Description
Combines high-visibilty work vest with a full-body harness that safely distributes fall arrest forces throughout the body.
- Vest is made of certified hi-vis fabric with reflective tape
- Delta No-Tangle™ harness design holds its shape, makes donning easy
- Highly durable polyester webbing has corrosion-resistant coated hardware
- Includes padded shoulders, neck liner, belt loops for a body belt, and convenient, Velcro™ tool fasteners for attaching accessories
- With two chest pockets, one with slots for pens and pencils, one with a snap closure to prevent loss of expensive items
Compliance
Meets or exceeds OSHA, ANSI Z359.1, and ANSI/ISEA 107–1999 standards.
Specifications
Specifications
| Color | Hi-vis Orange |
| D-Ring Location | Back |
| Size | Large Vest; Universal Harness |
| Certifications/Compliance | Meets or exceeds OSHA, ANSI Z359.1, and ANSIFor Use With (Application)/ISEA 107-1999 standards. |