Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ DBM 1285 dihydrochloride

Click to view available options
Quantity:
10 mg
50 mg
Description
DBM 1285 dihydrochloride is a p38 MAPK inhibitor. Supresses p38 phosphorylation and LPS-induced TNF-α production in macrophages and in vivo. Attenuates zymosan-induced inflammation and adjuvant-induced arthritis in murine models. Orally active.
Specifications
Specifications
| Quantity | 10 mg |
| Product Type | DBM 1285 dihydrochloride |
| Purity | >98% |
| Molecular Weight (g/mol) | 468.42 |
| Inhibitors | DBM 1285 dihydrochloride |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction