Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DBT Antibody (OTI1G2), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB270553A48 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB270553A48 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB270553A48 Supplier Novus Biologicals Supplier No. NBP270553AF488
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

DBT Monoclonal specifically detects DBT in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry.

Specifications

Antigen DBT
Applications Western Blot, Flow Cytometry
Classification Monoclonal
Clone OTI1G2
Conjugate Alexa Fluor 488
Dilution Western Blot, Flow Cytometry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias BCATE2BCKAD E2 subunit, BCKADE2, BCKAD-E2, branched chain acyltransferase, E2 component, Branched-chain alpha-keto acid dehydrogenase complex component E2, Dihydrolipoamide acetyltransferase component of branched-chain alpha-keto aciddehydrogenase complex, Dihydrolipoamide branched chain transacylase, dihydrolipoamide branched chain transacylase (E2 component of branched chainketo acid dehydrogenase complex; maple syrup urine disease), dihydrolipoamide branched chain transacylase E2, dihydrolipoyl transacylase, Dihydrolipoyllysine-residue (2-methylpropanoyl)transferase, E2, E2 component of branched chain alpha-keto acid dehydrogenase complex, E2B, EC 2.3.1.168, lipoamide acyltransferase component of branched-chain alpha-keto aciddehydrogenase complex, mitochondrial, lipoamide acyltransferase component of mitochondrial branched-chain alpha-ketoacid dehydrogenase complex, MGC9061, mitochondrial branched chain alpha-keto acid dehydrogenase transacylase subunit(E2b)
Gene Symbols DBT
Host Species Mouse
Immunogen Full length recombinant protein of human DBT (NP_001909) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 1629.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.