Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DBX1 Antibody, DyLight 650, Novus Biologicals™
SDP

Catalog No. NB163826 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB163826 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB163826 Supplier Novus Biologicals Supplier No. NBP281711C
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

DBX1 Polyclonal antibody specifically detects DBX1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen DBX1
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 650
Formulation 50mM Sodium Borate
Gene Alias developing brain homeobox 1, developing brain homeobox protein 1, homeobox protein DBX1
Host Species Rabbit
Immunogen DBX1 antibody was raised against a 15 amino acid synthetic peptide near the center of human DBX1. The immunogen is located within amino acids 100 - 150 of DBX1. Amino Acid Squence: AAILSSGPRTETSPAL
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 120237
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.