Learn More
Description
Specifications
Specifications
| Antigen | DC-LAMP |
| Applications | Flow Cytometry, Immunohistochemistry |
| Classification | Monoclonal |
| Clone | 1010E1.01 |
| Concentration | 0.5mg/mL |
| Conjugate | Alexa Fluor 647 |
| Dilution | Flow Cytometry 1:10-1:1000, Immunohistochemistry |
| Gene Accession No. | NP_055213 |
| Gene Alias | CD208 antigen, DC-LAMP, DCLAMPDC-lysosome-associated membrane glycoprotein, LAMP, LAMP-3, lysosomal-associated membrane protein 3DC LAMP, lysosome-associated membrane glycoprotein 3, Protein TSC403, TSC403CD208 |
| Gene Symbols | LAMP3 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
