Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DC-LAMP Antibody (109G3), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. DDX0193A647 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DDX0193A647 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. DDX0193A647 Supplier Novus Biologicals Supplier No. DDX0193A647100
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

DC-LAMP Monoclonal specifically detects DC-LAMP in Human samples. It is validated for Flow Cytometry, Immunohistochemistry.

Specifications

Antigen DC-LAMP
Applications Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone 109G3
Concentration 0.5mg/mL
Conjugate Alexa Fluor 647
Dilution Flow Cytometry 1:10-1:1000, Immunohistochemistry
Gene Accession No. NP_055213
Gene Alias CD208 antigen, DC-LAMP, DCLAMPDC-lysosome-associated membrane glycoprotein, LAMP, LAMP-3, lysosomal-associated membrane protein 3DC LAMP, lysosome-associated membrane glycoprotein 3, Protein TSC403, TSC403CD208
Gene Symbols LAMP3
Host Species Mouse
Immunogen DC-LAMP/CD208 was used as immunogen.
Purification Method Ion exchange chromatography
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Dendritic Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 27074
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.