Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DC-LAMP Antibody (L3P2H10*E3), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NBP250464AF Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP250464AF 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP250464AF Supplier Novus Biologicals Supplier No. NBP250464AF405
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

DC-LAMP Monoclonal specifically detects DC-LAMP in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.

Specifications

Antigen DC-LAMP
Applications Western Blot, ELISA, Immunohistochemistry
Classification Monoclonal
Clone L3P2H10*E3
Conjugate Alexa Fluor 405
Dilution Western Blot, ELISA, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias CD208 antigen, DC-LAMP, DCLAMPDC-lysosome-associated membrane glycoprotein, LAMP, LAMP-3, lysosomal-associated membrane protein 3DC LAMP, lysosome-associated membrane glycoprotein 3, Protein TSC403, TSC403CD208
Gene Symbols LAMP3
Host Species Mouse
Immunogen Ovalbumin-conjugated synthetic peptide - RCQSSGYQRI
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Dendritic Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 27074
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.