Learn More
DC-SIGN/CD209 Antibody (102E11.06), Alexa Fluor™ 647, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | DC-SIGN/CD209 |
| Applications | Flow Cytometry, Immunohistochemistry |
| Classification | Monoclonal |
| Clone | 102E11.06 |
| Concentration | 0.5mg/mL |
| Conjugate | Alexa Fluor 647 |
| Dilution | Flow Cytometry 1:10-1:1000, Immunohistochemistry |
| Gene Accession No. | NP_066978 |
| Gene Alias | CD209 antigendendritic cell-specific intracellular adhesion molecules (ICAM)-3 grabbingnon-integrin, CD209 molecule, CDSIGNHIV gpl20-binding protein, CLEC4LC-type lectin domain family 4 member L, DC-SIGN1C-type lectin domain family 4, member L, DC-SIGNMGC129965, Dendritic cell-specific ICAM-3-grabbing non-integrin 1 |
| Gene Symbols | CD209 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.